Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAGH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040427.1 | NP_001035517.1 | 5 | 839 | ||
NM_001286249.1 | NP_001273178.1 | 4 | 743 | |||
NM_005326.4 | NP_005317.2 | 4 | 743 | |||
XM_011522469.2 | XP_011520771.1 | 4 | 383 | |||
XM_011522470.2 | XP_011520772.1 | 4 | 381 | |||
XM_017023195.1 | XP_016878684.1 | 4 | 308 | |||
GenBank mRNA | AK290155.1 | - | 4 | 308 | ||
AK298174.1 | - | 4 | 383 | |||
AK299173.1 | - | 4 | 374 | |||
AK308825.1 | - | 4 | 351 | |||
BC000840.1 | - | 4 | 363 | 3457214 | ||
BC002627.2 | - | 4 | 692 | 3608098 | ||
BU176541.1 | - | 5 | 444 | |||
X90999.1 | - | 3 | 228 |