Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCLS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001292041.1 | NP_001278970.1 | 9 | 862 | ||
NM_005335.5 | NP_005326.2 | 10 | 973 | |||
GenBank mRNA | AK124015.1 | - | 5 | 1647 | ||
AK225633.1 | - | 9 | 846 | |||
AK298663.1 | - | 9 | 897 | |||
AK298696.1 | - | 9 | 812 | |||
AK312750.1 | - | 10 | 936 | |||
BC016758.2 | - | 10 | 893 | 2966303 | ||
BP338037.1 | - | 3 | 274 | |||
BT006824.1 | - | 9 | 844 | |||
BY994733.1 | - | 6 | 485 | |||
CR456794.1 | - | 9 | 844 | |||
X16663.1 | - | 10 | 886 |