Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HCRTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001526.3 | NP_001517.2 | 3 | 828 | ||
XM_017010798.1 | XP_016866287.1 | 3 | 783 | |||
GenBank mRNA | AF041245.1 | - | 3 | 872 | ||
AK307819.1 | - | 3 | 460 | |||
AK314279.1 | - | 3 | 780 | |||
KC812499.1 | - | 4 | 976 | |||
KC812500.1 | - | 4 | 976 |
Set Membership: |
G-protein coupled receptor |