Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001527.3 | NP_001518.3 | 8 | 1041 | ||
NR_033441.1 | 9 | 1069 | ||||
NR_073443.1 | 8 | 947 | ||||
XM_011535788.1 | XP_011534090.1 | 9 | 793 | |||
XM_017010799.1 | XP_016866288.1 | 9 | 1015 | |||
GenBank mRNA | AB209190.1 | - | 3 | 927 | ||
AK092156.1 | - | 8 | 947 | |||
AK097376.1 | - | 9 | 1003 | |||
AK296856.1 | - | 8 | 1041 | |||
BC031055.1 | - | 8 | 1005 | |||
U31814.1 | - | 8 | 958 |