Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDGF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126050.1 | NP_001119522.1 | 2 | 252 | ||
NM_001126051.1 | NP_001119523.1 | 2 | 227 | |||
NM_001319186.1 | NP_001306115.1 | 2 | 252 | |||
NM_001319187.1 | NP_001306116.1 | 2 | 77 | |||
NM_001319188.1 | NP_001306117.1 | 3 | 242 | |||
NM_004494.2 | NP_004485.1 | 2 | 441 | |||
NR_135008.1 | 2 | 252 | ||||
XM_011509453.2 | XP_011507755.2 | 2 | 264 | |||
XM_011509454.2 | XP_011507756.1 | 2 | 94 | |||
XM_011509455.2 | XP_011507757.1 | 2 | 160 | |||
GenBank mRNA | AK096411.1 | - | 2 | 228 | ||
AK292325.1 | - | 2 | 77 | |||
AK298342.1 | - | 2 | 191 | |||
AK304468.1 | - | 2 | 110 | |||
AK315514.1 | - | 2 | 210 | |||
AL833592.1 | - | 4 | 534 | |||
AU131311.1 | - | 2 | 256 | |||
BC018991.1 | - | 2 | 292 | 3503321 | ||
CA866443.1 | - | 2 | 86 | |||
CN392158.1 | - | 2 | 259 | |||
D16431.1 | - | 2 | 434 | |||
DA207052.1 | - | 2 | 114 | |||
DA367806.1 | - | 2 | 228 | |||
DB057555.1 | - | 3 | 242 | |||
DR159090.1 | - | 2 | 420 |