Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000186.3 | NP_000177.2 | 7 | 1109 | ||
NM_001014975.2 | NP_001014975.1 | 7 | 1109 | |||
XM_017001108.1 | XP_016856597.1 | 7 | 991 | |||
GenBank mRNA | AK225649.1 | - | 7 | 949 | ||
AK226113.1 | - | 2 | 2897 | |||
AK291395.1 | - | 7 | 948 | |||
BC012610.1 | - | 6 | 743 | |||
BC037285.2 | - | 7 | 934 | |||
BC110643.1 | - | 7 | 925 | |||
BC142699.1 | - | 7 | 921 | 40148771 | ||
M12383.1 | - | 2 | 194 | |||
X04697.1 | - | 6 | 713 | |||
X07523.1 | - | 7 | 942 | |||
Y00716.1 | - | 7 | 942 |