Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLA-DPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242524.1 | NP_001229453.1 | 4 | 764 | ||
NM_001242525.1 | NP_001229454.1 | 4 | 688 | |||
NM_033554.3 | NP_291032.2 | 3 | 673 | |||
GenBank mRNA | AK292709.1 | - | 3 | 580 | ||
BI553587.1 | - | 5 | 555 | |||
CD516536.1 | - | 4 | 696 | |||
M27487.1 | - | 3 | 601 | |||
X00457.1 | - | 3 | 486 |