Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002126.4 | NP_002117.1 | 3 | 1102 | ||
XM_005257269.2 | XP_005257326.1 | 3 | 1082 | |||
XM_011524705.1 | XP_011523007.1 | 3 | 556 | |||
XM_017024556.1 | XP_016880045.1 | 3 | 556 | |||
GenBank mRNA | AK074289.1 | - | 2 | 566 | ||
AK290043.1 | - | 3 | 539 | |||
AK295726.1 | - | 2 | 594 | |||
AK315079.1 | - | 3 | 740 | |||
BC036093.1 | - | 3 | 1056 | 5288156 | ||
CR541672.1 | - | 3 | 577 | |||
M95585.1 | - | 3 | 899 | |||
X68985.1 | - | 3 | 675 |