Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002126.4 | NP_002117.1 | 1, 2 | 637 | ||
XM_005257269.2 | XP_005257326.1 | 1, 2 | 617 | |||
GenBank mRNA | AK315079.1 | - | 1, 2 | 275 | ||
BC036093.1 | - | 1, 2 | 591 | 5288156 | ||
CR541672.1 | - | 1, 2 | 112 | |||
M95585.1 | - | 1, 2 | 434 | |||
X68985.1 | - | 1, 2 | 210 |