Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMBS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000190.3 | NP_000181.2 | 14 | 1216 | ||
NM_001024382.1 | NP_001019553.1 | 14 | 1118 | |||
NM_001258208.1 | NP_001245137.1 | 13 | 1096 | |||
NM_001258209.1 | NP_001245138.1 | 13 | 1187 | |||
XM_005271531.1 | XP_005271588.1 | 14 | 1294 | |||
XM_005271532.1 | XP_005271589.1 | 14 | 1272 | |||
XM_005271533.3 | XP_005271590.1 | 13 | 1168 | |||
XM_011542796.1 | XP_011541098.1 | 13 | 1548 | |||
XM_017017629.1 | XP_016873118.1 | 14 | 1682 | |||
GenBank mRNA | AK000628.1 | - | 13 | 1179 | ||
AK131072.1 | - | 13 | 1059 | |||
AK290275.1 | - | 14 | 1212 | |||
AW139390.1 | - | 1 | 307 | |||
BC000520.2 | - | 14 | 1210 | 2822949 | ||
BC008149.1 | - | 14 | 1171 | 4178944 | ||
BC019323.1 | - | 14 | 1210 | 2822949 | ||
BQ020524.1 | - | 1 | 308 | |||
BX647328.1 | - | 11 | 1821 | |||
X04217.1 | - | 14 | 1089 | |||
X04808.1 | - | 14 | 1086 |