Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGCS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098272.2 | NP_001091742.1 | 3 | 396 | ||
NM_001324219.1 | NP_001311148.1 | 2 | 383 | |||
NM_001324220.1 | NP_001311149.1 | 3 | 416 | |||
NM_001324222.1 | NP_001311151.1 | 3 | 396 | |||
NM_001324223.1 | NP_001311152.1 | 2 | 337 | |||
NM_001324224.1 | NP_001311153.1 | 3 | 416 | |||
NM_002130.7 | NP_002121.4 | 2 | 337 | |||
XM_017009412.1 | XP_016864901.1 | 3 | 476 | |||
GenBank mRNA | AK095492.1 | - | 2 | 237 | ||
AK315593.1 | - | 3 | 324 | |||
BC000297.2 | - | 3 | 275 | |||
BC083514.1 | - | 2 | 235 | 4443624 | ||
BJ995153.1 | - | 3 | 380 | |||
BT007302.1 | - | 1 | 170 | |||
DA801434.1 | - | 2 | 384 | |||
DB074885.1 | - | 2 | 338 | |||
DB078662.1 | - | 2 | 318 | |||
L25798.1 | - | 1 | 191 | |||
X66435.1 | - | 3 | 292 |