Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127204.1 | NP_001120676.1 | 4 | 495 | ||
NM_001127205.1 | NP_001120677.1 | 3 | 399 | |||
NM_001127206.2 | NP_001120678.1 | 3 | 374 | |||
NM_001286267.1 | NP_001273196.1 | 4 | 498 | |||
NM_001286268.1 | NP_001273197.1 | 3 | 555 | |||
NM_001286269.1 | NP_001273198.1 | 4 | 476 | |||
NM_001286270.1 | NP_001273199.1 | 5 | 775 | |||
NM_001286271.1 | NP_001273200.1 | 2 | 250 | |||
NM_002134.3 | NP_002125.3 | 3 | 377 | |||
XM_011522473.1 | XP_011520775.1 | 3 | 365 | |||
XM_017023196.1 | XP_016878685.1 | 4 | 419 | |||
XM_017023197.1 | XP_016878686.1 | 3 | 250 | |||
GenBank mRNA | AF051306.1 | - | 2 | 162 | ||
AK093388.1 | - | 5 | 736 | |||
AK222629.1 | - | 3 | 269 | |||
AK313660.1 | - | 3 | 329 | |||
BC002396.2 | - | 3 | 293 | 2821444 | ||
BE561721.1 | - | 3 | 411 | |||
BI225653.1 | - | 3 | 376 | |||
BM905872.1 | - | 2 | 155 | |||
BP329113.1 | - | 4 | 377 | |||
BP368949.1 | - | 3 | 399 | |||
BP369861.1 | - | 3 | 337 | |||
BT019788.1 | - | 2 | 188 | |||
CR456760.1 | - | 2 | 188 | |||
CR541839.1 | - | 2 | 188 | |||
D21243.1 | - | 2 | 188 | |||
DA141643.1 | - | 4 | 498 | |||
DA694137.1 | - | 4 | 424 | |||
DB184091.1 | - | 3 | 483 | |||
S34389.1 | - | 3 | 272 |