Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPA2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002137.3 | NP_002128.1 | 9 | 1145 | ||
NM_031243.2 | NP_112533.1 | 10 | 1181 | |||
XM_005249729.1 | XP_005249786.1 | 10 | 1163 | |||
XM_017012109.1 | XP_016867598.1 | 9 | 1129 | |||
XM_017012110.1 | XP_016867599.1 | 9 | 1129 | |||
GenBank mRNA | AK129839.1 | - | 9 | 1099 | ||
AK289429.1 | - | 10 | 1134 | |||
BC000506.2 | - | 8 | 913 | 2822109 | ||
BX537494.1 | - | 8 | 3978 | |||
BX641084.1 | - | 8 | 4719 | |||
M29064.1 | - | 10 | 1114 | |||
M29065.1 | - | 9 | 1084 |