Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPA2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002137.3 | NP_002128.1 | 4 | 562 | ||
NM_031243.2 | NP_112533.1 | 5 | 598 | |||
XM_005249729.1 | XP_005249786.1 | 5 | 580 | |||
XM_017012109.1 | XP_016867598.1 | 4 | 546 | |||
XM_017012110.1 | XP_016867599.1 | 4 | 546 | |||
GenBank mRNA | AK129839.1 | - | 4 | 516 | ||
AK289429.1 | - | 5 | 551 | |||
BC000506.2 | - | 4 | 493 | 2822109 | ||
BX537494.1 | - | 3 | 3395 | |||
BX641084.1 | - | 3 | 4135 | |||
M29064.1 | - | 5 | 531 | |||
M29065.1 | - | 4 | 501 |