Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPA2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002137.3 | NP_002128.1 | 5 | 734 | ||
NM_031243.2 | NP_112533.1 | 6 | 770 | |||
XM_005249729.1 | XP_005249786.1 | 6 | 752 | |||
XM_017012109.1 | XP_016867598.1 | 5 | 718 | |||
XM_017012110.1 | XP_016867599.1 | 5 | 718 | |||
GenBank mRNA | AK129839.1 | - | 5 | 688 | ||
AK289429.1 | - | 6 | 723 | |||
BC000506.2 | - | 5 | 665 | 2822109 | ||
BX537494.1 | - | 4 | 3567 | |||
BX641084.1 | - | 4 | 4307 | |||
M29064.1 | - | 6 | 703 | |||
M29065.1 | - | 5 | 673 |