Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004499.3 | NP_004490.2 | 3 | 547 | ||
NM_031266.2 | NP_112556.2 | 3 | 547 | |||
GenBank mRNA | AK054600.1 | - | 2 | 595 | ||
AK097657.1 | - | 3 | 548 | |||
AK223425.1 | - | 3 | 524 | |||
BC001616.1 | - | 3 | 506 | 3547193 | ||
BC002625.2 | - | 3 | 506 | 3606731 | ||
BC004561.2 | - | 3 | 514 | 3960088 | ||
BC009359.2 | - | 3 | 510 | 4301401 | ||
BC036708.2 | - | 3 | 473 | 4331453 | ||
M65028.1 | - | 3 | 426 | |||
U76713.1 | - | 2 | 299 |