Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098204.1 | NP_001091674.1 | 4 | 1404 | ||
NM_001098205.1 | NP_001091675.1 | 4 | 1444 | |||
NM_001098206.1 | NP_001091676.1 | 4 | 1421 | |||
NM_001098207.1 | NP_001091677.1 | 4 | 1448 | |||
NM_001098208.1 | NP_001091678.1 | 3 | 1527 | |||
NM_004966.3 | NP_004957.1 | 4 | 1405 | |||
GenBank mRNA | AB062287.1 | - | 1 | 108 | ||
AK001364.1 | - | 4 | 1442 | |||
AK296696.1 | - | 5 | 1225 | |||
BC001432.2 | - | 3 | 1199 | 3138435 | ||
BC004254.1 | - | 4 | 1363 | 3633625 | ||
BC015580.2 | - | 4 | 1320 | 4636807 | ||
BC016736.1 | - | 4 | 1393 | 4104978 | ||
BC106008.1 | - | 4 | 1332 | 6156823 | ||
L28010.1 | - | 1 | 1068 |