Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HNRNPH1 SIRNA Assays ›
Gene Symbol:
HNRNPH1
Gene Name:
heterogeneous nuclear ribonucleoprotein H1 (H)
Gene Aliases:
HNRPH, HNRPH1, hnRNPH
Chromosome Location:
Chr.5: 179614178 - 179634784 on Build GRCh38
UniGene ID:
Hs.604001
Species:
Human
siRNA ID
HSS104907
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HNRNPH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001257293.1
NP_001244222.1
4
512
NM_005520.2
NP_005511.1
4
482
XM_005265895.2
XP_005265952.1
4
469
XM_005265896.2
XP_005265953.1
4
472
XM_005265901.1
XP_005265958.1
4
473
XM_005265902.4
XP_005265959.1
4
459
XM_006714862.2
XP_006714925.1
4
472
XM_006714863.2
XP_006714926.1
4
472
XM_011534542.1
XP_011532844.1
4
786
XM_011534544.1
XP_011532846.1
4
969
XM_011534545.1
XP_011532847.1
4
677
XM_011534546.1
XP_011532848.1
5
825
XM_011534547.1
XP_011532849.1
4
472
XM_017009414.1
XP_016864903.1
4
469
XM_017009415.1
XP_016864904.1
4
491
XM_017009416.1
XP_016864905.1
4
786
XM_017009417.1
XP_016864906.1
5
825
XM_017009418.1
XP_016864907.1
4
472
XM_017009419.1
XP_016864908.1
4
469
XM_017009420.1
XP_016864909.1
4
472
XM_017009421.1
XP_016864910.1
4
472
XM_017009422.1
XP_016864911.1
4
459
XM_017009423.1
XP_016864912.1
4
472
XM_017009424.1
XP_016864913.1
3
697
XM_017009425.1
XP_016864914.1
4
472
GenBank mRNA
AK092273.1
-
3
679
AK124530.1
-
3
1901
AK307693.1
-
4
473
BC001348.2
-
4
461
2961378
CR456778.1
-
3
365
DA204744.1
-
4
512
L22009.1
-
4
437
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
RNA processing
fibroblast growth factor receptor signaling pathway
gene expression
regulation of RNA splicing
nucleotide binding
RNA binding
protein binding
poly(U) RNA binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-67fd5c5bdd-gnqd7:80/10.239.161.169:80.
git-commit: 7fb76fe7d0d673187a4c374cd16a9abd242c4866
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.3-offline