Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HNRNPU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004501.3 | NP_004492.2 | 2 | 920 | ||
NM_031844.2 | NP_114032.2 | 2 | 977 | |||
XM_017001115.1 | XP_016856604.1 | 2 | 1172 | |||
XM_017001116.1 | XP_016856605.1 | 2 | 977 | |||
XM_017001117.1 | XP_016856606.1 | 2 | 966 | |||
GenBank mRNA | AF068846.1 | - | 2 | 973 | ||
AK126868.1 | - | 3 | 718 | |||
AK130669.1 | - | 1 | 26 | |||
AK297119.1 | - | 3 | 811 | |||
AK307661.1 | - | 2 | 444 | |||
BC003367.1 | - | 2 | 851 | 2900876 | ||
BC003621.2 | - | 2 | 853 | 2966453 | ||
BC007950.2 | - | 2 | 401 | |||
BC015782.1 | - | 2 | 454 | 4857112 | ||
BC024767.1 | - | 2 | 853 | 4993703 | ||
X65488.1 | - | 2 | 743 |