Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004502.3 | NP_004493.3 | 2 | 634 | ||
GenBank mRNA | AK223249.1 | - | 2 | 633 | ||
AK290653.1 | - | 2 | 600 | |||
BC015345.1 | - | 2 | 581 | 4413080 | ||
BM923808.1 | - | 2 | 635 | |||
M16937.1 | - | 2 | 630 | |||
M30598.1 | - | 2 | 177 | |||
S49765.1 | - | 2 | 240 |