Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024016.3 | NP_076921.1 | 1 | 306 | ||
XM_005257286.2 | XP_005257343.1 | 3 | 5898 | |||
XM_017024564.1 | XP_016880053.1 | 4 | 5622 | |||
GenBank mRNA | AK096222.1 | - | 1 | 306 |