Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024501.2 | NP_078777.1 | 2 | 925 | ||
XM_005246507.3 | XP_005246564.1 | 3 | 966 | |||
GenBank mRNA | AF241528.1 | - | 2 | 925 | ||
AK314120.1 | - | 2 | 840 | |||
BC014477.1 | - | 2 | 829 | 4869019 | ||
BC028190.1 | - | 5 | 653 |