Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006898.4 | NP_008829.3 | 2 | 230 | ||
XM_005246509.3 | XP_005246566.1 | 3 | 497 | |||
XM_005246510.4 | XP_005246567.1 | 3 | 313 | |||
XM_005246511.3 | XP_005246568.1 | 3 | 921 | |||
XM_005246513.4 | XP_005246570.1 | 3 | 524 | |||
XM_006712477.2 | XP_006712540.1 | 2 | 207 | |||
XM_011511065.2 | XP_011509367.1 | 3 | 2301 | |||
XM_011511066.2 | XP_011509368.1 | 3 | 375 | |||
GenBank mRNA | BC005124.2 | - | 2 | 231 | 3936607 | |
BC008789.2 | - | 1 | 162 | 3628279 |