Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002148.3 | NP_002139.2 | 2 | 967 | ||
GenBank mRNA | AW243295.1 | - | 1 | 591 | ||
BC069619.1 | - | 2 | 947 | 7262455 | ||
BC074760.2 | - | 2 | 946 | 30915219 | ||
X59373.1 | - | 2 | 930 |