Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171993.1 | NP_001165464.1 | 12 | 1198 | ||
NM_002150.2 | NP_002141.1 | 10 | 722 | |||
GenBank mRNA | AK057510.1 | - | 12 | 1198 | ||
AK290826.1 | - | 10 | 718 | |||
BC024287.1 | - | 10 | 704 | 5087393 | ||
X72389.1 | - | 10 | 707 |