Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD11B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206741.1 | NP_001193670.1 | 6 | 689 | ||
NM_005525.3 | NP_005516.1 | 5 | 687 | |||
NM_181755.2 | NP_861420.1 | 6 | 667 | |||
GenBank mRNA | AK313973.1 | - | 5 | 629 | ||
BC012593.1 | - | 5 | 636 | 4286028 | ||
BX345837.2 | - | 5 | 600 | |||
KJ534869.1 | - | 5 | 535 |