Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021979.3 | NP_068814.2 | 1 | 1842 | ||
GenBank mRNA | AK097824.1 | - | 2 | 891 | ||
BC001752.1 | - | 2 | 1536 | 3358246 | ||
BC036107.1 | - | 1 | 1568 | 5276121 | ||
BT009815.1 | - | 1 | 1460 |