Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006597.5 | NP_006588.1 | 5 | 1025 | ||
NM_153201.3 | NP_694881.1 | 5 | 1025 | |||
XM_011542798.1 | XP_011541100.1 | 5 | 773 | |||
GenBank mRNA | AB034951.1 | - | 5 | 789 | ||
AB073886.1 | - | 4 | 703 | |||
AF352832.1 | - | 5 | 812 | |||
AK096100.1 | - | 5 | 751 | |||
AK129885.1 | - | 5 | 827 | |||
AK222628.1 | - | 5 | 823 | |||
AK222785.1 | - | 5 | 823 | |||
AK292067.1 | - | 2 | 455 | |||
AK300404.1 | - | 4 | 986 | |||
AK303935.1 | - | 4 | 596 | |||
AK310467.1 | - | 5 | 897 | |||
BC007276.1 | - | 4 | 573 | |||
BC016179.1 | - | 5 | 806 | 3920744 | ||
BC016660.1 | - | 5 | 806 | 3923981 | ||
BC019816.2 | - | 5 | 806 | 2899894 | ||
EU668349.1 | - | 5 | 806 | |||
FJ224294.1 | - | 5 | 826 |