Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300914.1 | NP_001287843.1 | 2 | 646 | ||
NM_001313964.1 | NP_001300893.1 | 3 | 572 | |||
NM_006145.2 | NP_006136.1 | 2 | 795 | |||
XM_006722733.1 | XP_006722796.1 | 3 | 585 | |||
XM_006722734.3 | XP_006722797.1 | 3 | 1009 | |||
XM_011527956.2 | XP_011526258.1 | 3 | 564 | |||
GenBank mRNA | AK092989.1 | - | 1 | 198 | ||
AK301817.1 | - | 3 | 487 | |||
AK312624.1 | - | 2 | 656 | |||
BC002352.2 | - | 2 | 619 | 2821008 | ||
BC019827.1 | - | 2 | 609 | 4152608 | ||
CR536485.1 | - | 2 | 585 | |||
CR541677.1 | - | 2 | 585 | |||
D49547.1 | - | 2 | 625 | |||
X62421.1 | - | 2 | 630 |