Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291860.1 | NP_001278789.1 | 48 | 6201 | ||
NM_005529.6 | NP_005520.4 | 48 | 6198 | |||
XM_011541318.2 | XP_011539620.1 | 53 | 6682 | |||
XM_017001120.1 | XP_016856609.1 | 49 | 6328 | |||
XM_017001121.1 | XP_016856610.1 | 48 | 6277 | |||
XM_017001122.1 | XP_016856611.1 | 48 | 6274 | |||
GenBank mRNA | X62515.1 | - | 48 | 6164 |