Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NDST1 SIRNA Assays ›
Gene Symbol:
NDST1
Gene Name:
N-deacetylase and N-sulfotransferase 1
Gene Aliases:
HSST, MRT46, NST1
Chromosome Location:
Chr.5: 150485783 - 150558211 on Build GRCh38
UniGene ID:
Hs.222055
Species:
Human
siRNA ID
HSS105105
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NDST1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001301063.1
NP_001287992.1
14
2998
NM_001543.4
NP_001534.1
15
3082
XM_005268433.1
XP_005268490.1
15
3093
XM_005268434.2
XP_005268491.1
15
3702
XM_005268435.4
XP_005268492.1
15
3227
XM_005268436.3
XP_005268493.1
15
4867
XM_005268437.4
XP_005268494.1
15
3106
XM_005268438.1
XP_005268495.1
14
2922
XM_005268439.1
XP_005268496.1
14
2883
XM_005268442.3
XP_005268499.1
13
2076
XM_006714782.1
XP_006714845.1
15
3128
XM_006714783.1
XP_006714846.1
15
3054
XM_011537638.2
XP_011535940.1
14
2183
XM_017009427.1
XP_016864916.1
15
6084
XM_017009428.1
XP_016864917.1
15
3187
XM_017009429.1
XP_016864918.1
15
3067
XM_017009430.1
XP_016864919.1
15
3089
XM_017009431.1
XP_016864920.1
15
3659
XM_017009432.1
XP_016864921.1
14
4879
XM_017009433.1
XP_016864922.1
14
4838
GenBank mRNA
AB209107.1
-
15
2856
AK292448.1
-
15
3082
AK293746.1
-
14
2998
U17970.1
-
14
2808
U18918.1
-
14
2796
U36600.1
-
15
3003
Back To Top
More Information
Panther Classification:
Molecular Function -
transferase
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
polysaccharide biosynthetic process
cardiac septum development
glycosaminoglycan biosynthetic process
protein sulfation
inflammatory response
smoothened signaling pathway
respiratory gaseous exchange
fibroblast growth factor receptor signaling pathway
heparan sulfate proteoglycan biosynthetic process
heparin biosynthetic process
forebrain development
midbrain development
aorta development
embryonic neurocranium morphogenesis
embryonic viscerocranium morphogenesis
coronary vasculature development
protein binding
[heparan sulfate]-glucosamine N-sulfotransferase activity
N-acetylglucosamine deacetylase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-hnnbr:80/10.239.164.27:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline