Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000869.5 | NP_000860.2 | 4 | 556 | ||
NM_001161772.2 | NP_001155244.1 | 4 | 315 | |||
NM_213621.3 | NP_998786.2 | 4 | 556 | |||
NR_046363.1 | 4 | 556 | ||||
GenBank mRNA | AF498984.1 | - | 4 | 305 | ||
AJ003078.1 | - | 4 | 345 | |||
AJ003079.1 | - | 4 | 345 | |||
AJ003080.1 | - | 4 | 345 | |||
AK299973.1 | - | 4 | 315 | |||
AK304630.1 | - | 4 | 558 | |||
AK313896.1 | - | 4 | 435 | |||
BC002354.2 | - | 4 | 372 | 2821710 | ||
BC004453.1 | - | 4 | 424 | 2819400 | ||
BT007204.1 | - | 4 | 323 | |||
CD014118.1 | - | 4 | 443 | |||
D49394.1 | - | 4 | 524 | |||
S82612.1 | - | 4 | 347 |