Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000872.4 | NP_000863.1 | 2 | 1449 | ||
NM_019859.3 | NP_062873.1 | 2 | 1449 | |||
NM_019860.3 | NP_062874.1 | 2 | 1449 | |||
GenBank mRNA | AB451482.1 | - | 2 | 1206 | ||
AK292606.1 | - | 2 | 1219 | |||
AY493988.1 | - | 2 | 1206 | |||
BC047526.1 | - | 2 | 1449 | 5297325 | ||
L21195.1 | - | 2 | 1233 | |||
U68487.1 | - | 2 | 1233 | |||
U68488.1 | - | 2 | 1233 |
Set Membership: |
G-protein coupled receptor |