Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002161.5 | NP_002152.2 | 28 | 3126 | ||
NM_013417.3 | NP_038203.2 | 28 | 3296 | |||
NR_073446.1 | 27 | 2969 | ||||
GenBank mRNA | AB209234.1 | - | 28 | 3035 | ||
AK293014.1 | - | 28 | 3053 | |||
AK301272.1 | - | 16 | 1606 | |||
AK308202.1 | - | 9 | 862 | |||
BC008318.1 | - | 6 | 631 | |||
BC065552.1 | - | 27 | 2968 | 6043344 | ||
BX537417.1 | - | 28 | 3064 | |||
BX537429.1 | - | 28 | 3066 | |||
BX538109.1 | - | 27 | 2909 | |||
BX647071.1 | - | 28 | 3065 | |||
BX647236.1 | - | 28 | 3058 | |||
BX647430.1 | - | 28 | 3060 | |||
BX648567.1 | - | 28 | 3066 | |||
D28473.1 | - | 28 | 3214 | |||
U04953.1 | - | 28 | 3228 |