Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002166.4 | NP_002157.2 | 1 | 285 | ||
GenBank mRNA | AK222682.1 | - | 1 | 212 | ||
AK311988.1 | - | 1 | 212 | |||
AY634687.1 | - | 1 | 221 | |||
BC030639.2 | - | 1 | 212 | 4820416 | ||
BE222494.1 | - | 1 | 285 | |||
D13891.1 | - | 1 | 198 | |||
M97796.1 | - | 1 | 210 |