Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165946.1 | NP_001159418.1 | 11, 12 | 1429 | ||
NM_001322793.1 | NP_001309722.1 | 21, 22 | 2845 | |||
NM_001322794.1 | NP_001309723.1 | 21, 22 | 2728 | |||
NM_001322795.1 | NP_001309724.1 | 22, 23 | 3027 | |||
NM_001322796.1 | NP_001309725.1 | 21, 22 | 3009 | |||
NM_001322797.1 | NP_001309726.1 | 11, 12 | 1429 | |||
NM_004969.3 | NP_004960.2 | 21, 22 | 2845 | |||
NR_136399.1 | 22, 23 | 2969 | ||||
XM_017016187.1 | XP_016871676.1 | 21, 22 | 2768 | |||
XM_017016188.1 | XP_016871677.1 | 21, 22 | 3362 | |||
XM_017016189.1 | XP_016871678.1 | 22, 23 | 2761 | |||
XM_017016190.1 | XP_016871679.1 | 21, 22 | 2888 | |||
GenBank mRNA | AB209204.1 | - | 12, 13 | 1494 | ||
AK093287.1 | - | 11, 12 | 1401 | |||
AK302578.1 | - | 11, 12 | 1382 | |||
AK312810.1 | - | 21, 22 | 2770 | |||
AK316407.1 | - | 11, 12 | 1401 | |||
BC096336.1 | - | 21, 22 | 2809 | 40008464 | ||
BC096337.1 | - | 21, 22 | 2809 | 40008465 | ||
BC096339.3 | - | 21, 22 | 2809 | 40008468 | ||
BX648462.1 | - | 21, 22 | 3010 | |||
M21188.1 | - | 21, 22 | 2826 |