Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282386.1 | NP_001269315.1 | 4, 5 | 623 | ||
NM_001282387.1 | NP_001269316.1 | 4, 5 | 780 | |||
NM_005896.3 | NP_005887.2 | 4, 5 | 712 | |||
GenBank mRNA | AF020038.1 | - | 4, 5 | 666 | ||
AF113917.1 | - | 4, 5 | 603 | |||
AK302105.1 | - | 2, 3 | 158 | |||
AK312242.1 | - | 4, 5 | 683 | |||
AL136702.1 | - | 4, 5 | 644 | |||
BC012846.1 | - | 4, 5 | 663 | 3880331 | ||
BC021046.1 | - | 4, 5 | 609 | |||
BC093020.1 | - | 4, 5 | 641 | 30528912 | ||
BX537411.1 | - | 4, 5 | 612 | |||
CR533522.1 | - | 2, 3 | 417 | |||
CR536499.1 | - | 2, 3 | 417 | |||
CR541695.1 | - | 2, 3 | 417 | |||
DQ403082.1 | - | 2, 3 | 330 | |||
DR158630.1 | - | 4, 5 | 780 | |||
EU794623.1 | - | 4, 5 | 651 | |||
FJ224342.1 | - | 4, 5 | 612 | |||
U62389.1 | - | 1, 2 | 121 |