Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IDH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289910.1 | NP_001276839.1 | 2 | 117 | ||
NM_002168.3 | NP_002159.2 | 2 | 350 | |||
GenBank mRNA | AK222916.1 | - | 2 | 264 | ||
AK294148.1 | - | 2 | 117 | |||
AK312627.1 | - | 2 | 314 | |||
AK316388.1 | - | 2 | 191 | |||
BC009244.2 | - | 2 | 252 | 2959540 | ||
BC071828.1 | - | 2 | 251 | 6090792 | ||
X69433.1 | - | 2 | 272 |