Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206567.1 | NP_001193496.1 | 9 | 2156 | ||
NM_005531.2 | NP_005522.2 | 9 | 2156 | |||
XM_005245127.3 | XP_005245184.1 | 11 | 2641 | |||
XM_006711290.1 | XP_006711353.1 | 8 | 1988 | |||
XM_017001149.1 | XP_016856638.1 | 9 | 2305 | |||
GenBank mRNA | AB208989.1 | - | 9 | 2126 | ||
AF208043.1 | - | 9 | 2130 | |||
AK094968.1 | - | 7 | 1529 | |||
AK296228.1 | - | 9 | 2039 | |||
AK307957.1 | - | 9 | 1937 | |||
BC017059.1 | - | 9 | 2123 | 3914632 | ||
M63838.1 | - | 9 | 2130 |