Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFALS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146006.1 | NP_001139478.1 | 2 | 1743 | ||
NM_004970.2 | NP_004961.1 | 2 | 1629 | |||
NR_027389.1 | 2 | 1602 | ||||
GenBank mRNA | AK292182.1 | - | 2 | 1596 | ||
AK303146.1 | - | 2 | 1742 | |||
AK313198.1 | - | 2 | 1596 | |||
AK315808.1 | - | 2 | 1622 | |||
BC025681.1 | - | 2 | 1629 | 5205012 | ||
M86826.1 | - | 2 | 1604 |