Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000586.3 | NP_000577.2 | 4 | 471 | ||
GenBank mRNA | A14844.1 | - | 4 | 426 | ||
AB451325.1 | - | 4 | 416 | |||
AB451454.1 | - | 4 | 416 | |||
AF228636.1 | - | 4 | 428 | |||
AY283686.1 | - | 4 | 356 | |||
AY523040.1 | - | 4 | 356 | |||
BC066254.1 | - | 4 | 434 | 6971690 | ||
BC066255.1 | - | 4 | 434 | 6971691 | ||
BC066256.1 | - | 4 | 434 | 6971693 | ||
BC066257.1 | - | 4 | 434 | 6971694 | ||
BC070338.1 | - | 4 | 463 | 30418817 | ||
DQ231169.1 | - | 4 | 356 | |||
DQ861285.1 | - | 4 | 418 | |||
S77834.1 | - | 4 | 463 | |||
S82692.1 | - | 4 | 710 | |||
U25676.1 | - | 4 | 472 | |||
V00564.1 | - | 4 | 463 | |||
X01586.1 | - | 4 | 523 |