Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL2RB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000878.3 | NP_000869.1 | 8 | 908 | ||
GenBank mRNA | AK312860.1 | - | 8 | 902 | ||
BC025691.1 | - | 8 | 891 | 5207833 | ||
CR456506.1 | - | 7 | 796 | |||
M26062.1 | - | 8 | 908 |