Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL10RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001558.3 | NP_001549.2 | 7 | 1430 | ||
NR_026691.1 | 8 | 1560 | ||||
GenBank mRNA | AB209626.1 | - | 6 | 2931 | ||
AK291645.1 | - | 7 | 1430 | |||
AK301260.1 | - | 6 | 1248 | |||
AK301484.1 | - | 8 | 1560 | |||
AK301492.1 | - | 7 | 1700 | |||
AK313049.1 | - | 7 | 1424 | |||
AK316146.1 | - | 7 | 1482 | |||
AK316342.1 | - | 8 | 1634 | |||
BC028082.1 | - | 7 | 1407 | 5216986 | ||
JN591377.1 | - | 7 | 1353 | |||
U00672.1 | - | 7 | 1414 |