Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL10RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001558.3 | NP_001549.2 | 7 | 1569 | ||
NR_026691.1 | 8 | 1699 | ||||
GenBank mRNA | AB209626.1 | - | 6 | 3070 | ||
AK291645.1 | - | 7 | 1569 | |||
AK301260.1 | - | 6 | 1387 | |||
AK301484.1 | - | 8 | 1699 | |||
AK301492.1 | - | 7 | 1839 | |||
AK313049.1 | - | 7 | 1563 | |||
AK316146.1 | - | 7 | 1621 | |||
AK316342.1 | - | 8 | 1773 | |||
BC028082.1 | - | 7 | 1546 | 5216986 | ||
JN591377.1 | - | 7 | 1492 | |||
U00672.1 | - | 7 | 1553 |