Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL13RA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001560.2 | NP_001551.1 | 10 | 1182 | ||
XM_011531336.1 | XP_011529638.1 | 10 | 1460 | |||
XM_011531337.1 | XP_011529639.1 | 3 | 1489 | |||
XM_017029507.1 | XP_016884996.1 | 9 | 1199 | |||
GenBank mRNA | AB209849.1 | - | 10 | 1137 | ||
AK123515.1 | - | 3 | 1145 | |||
AK296958.1 | - | 5 | 411 | |||
AK313467.1 | - | 10 | 1171 | |||
BC009960.2 | - | 10 | 1169 | 4300487 | ||
BC015768.1 | - | 10 | 1186 | 4868206 | ||
U81379.3 | - | 10 | 1179 | |||
Y09328.1 | - | 10 | 1172 | |||
Y10659.1 | - | 10 | 1182 |