Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL13RA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001560.2 | NP_001551.1 | 7, 8 | 917 | ||
XM_011531336.1 | XP_011529638.1 | 7, 8 | 1195 | |||
XM_017029507.1 | XP_016884996.1 | 6, 7 | 934 | |||
GenBank mRNA | AB209849.1 | - | 7, 8 | 872 | ||
AK296958.1 | - | 2, 3 | 146 | |||
AK313467.1 | - | 7, 8 | 906 | |||
BC009960.2 | - | 7, 8 | 904 | 4300487 | ||
BC015768.1 | - | 7, 8 | 921 | 4868206 | ||
U81379.3 | - | 7, 8 | 914 | |||
Y09328.1 | - | 7, 8 | 907 | |||
Y10659.1 | - | 7, 8 | 917 |