Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL13RA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000640.2 | NP_000631.1 | 5 | 610 | ||
GenBank mRNA | AK291957.1 | - | 5 | 579 | ||
AK308523.1 | - | 5 | 541 | |||
AK309444.1 | - | 5 | 620 | |||
BC020739.1 | - | 4 | 511 | 4807603 | ||
BC033705.2 | - | 4 | 519 | 3876064 | ||
GQ494004.1 | - | 5 | 542 | |||
U70981.1 | - | 5 | 578 | |||
X95302.1 | - | 5 | 537 | |||
Y08768.1 | - | 5 | 610 |