Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL15RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243539.1 | NP_001230468.1 | 7 | 1088 | ||
NM_001256765.1 | NP_001243694.1 | 8 | 1037 | |||
NM_002189.3 | NP_002180.1 | 7 | 884 | |||
NM_172200.2 | NP_751950.2 | 6 | 785 | |||
NR_046362.1 | 5 | 794 | ||||
XM_011519465.1 | XP_011517767.1 | 9 | 1278 | |||
XM_011519466.2 | XP_011517768.1 | 9 | 1398 | |||
XM_011519469.1 | XP_011517771.1 | 8 | 1179 | |||
XM_011519476.2 | XP_011517778.1 | 5 | 739 | |||
XM_011519477.2 | XP_011517779.1 | 5 | 864 | |||
XM_017016195.1 | XP_016871684.1 | 8 | 1125 | |||
XM_017016198.1 | XP_016871687.1 | 7 | 1009 | |||
XM_017016201.1 | XP_016871690.1 | 7 | 1253 | |||
XM_017016202.1 | XP_016871691.1 | 8 | 974 | |||
XM_017016203.1 | XP_016871692.1 | 6 | 1436 | |||
GenBank mRNA | AF035279.1 | - | 7 | 819 | ||
AK304211.1 | - | 6 | 696 | |||
BC065506.1 | - | 5 | 786 | |||
BC074726.2 | - | 7 | 796 | 30915179 | ||
BC107777.1 | - | 7 | 952 | 6497994 | ||
BC121140.2 | - | 7 | 791 | 40120998 | ||
BC121141.2 | - | 7 | 796 | 40120999 | ||
CR457064.1 | - | 6 | 657 | |||
CR542023.1 | - | 7 | 765 | |||
HQ401283.1 | - | 8 | 918 | |||
U31628.1 | - | 7 | 847 |