Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ILF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267809.1 | NP_001254738.1 | 13 | 1089 | ||
NM_004515.3 | NP_004506.2 | 13 | 1070 | |||
GenBank mRNA | AK223326.1 | - | 13 | 990 | ||
AK302214.1 | - | 13 | 1033 | |||
AK313364.1 | - | 13 | 1029 | |||
BC000382.2 | - | 13 | 995 | 2820505 | ||
HQ322261.1 | - | 12 | 964 | |||
HQ322262.1 | - | 12 | 964 | |||
U10323.1 | - | 13 | 1003 |