Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014214.2 | NP_055029.1 | 2 | 421 | ||
XM_011525661.2 | XP_011523963.1 | 2 | 384 | |||
GenBank mRNA | AF014398.2 | - | 2 | 421 | ||
AF200432.1 | - | 1 | 98 | |||
BC017176.1 | - | 2 | 261 | 3345789 | ||
BC031560.1 | - | 2 | 333 | |||
BT007061.1 | - | 2 | 179 |